Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromomethyl methyl ether, 90%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
13057-17-5
C2H5BrO
124.97
JAMFGQBENKSWOF-UHFFFAOYSA-N
bromomethyl methyl ether,methane, bromomethoxy,bromo methoxy methane,methoxymethyl bromide,ether, bromomethyl methyl,bromomethylmethylether,methoxymethylbromide,mombr,bromo-methoxy-methane,bromomethyl methylether
83093
bromo(methoxy)methane
COCBr
13057-17-5
C2H5BrO
124.97
JAMFGQBENKSWOF-UHFFFAOYSA-N
bromomethyl methyl ether,methane, bromomethoxy,bromo methoxy methane,methoxymethyl bromide,ether, bromomethyl methyl,bromomethylmethylether,methoxymethylbromide,mombr,bromo-methoxy-methane,bromomethyl methylether
83093
bromo(methoxy)methane
COCBr
Quantity 5g
Boiling Point 87.0°C
Density 1.5300g/mL
Packaging Glass bottle
Flash Point 26°C
Specific Gravity 1.53
Refractive Index 1.455
Assay Percent Range 88% min. (GC)
Linear Formula BrCH2OCH3
Beilstein 01,582
Fieser 10,56
Formula Weight 124.97
Percent Purity 90%
Chemical Name or Material Bromomethyl methyl ether
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a P

GHS Signal Word: Warning