Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bromopentacarbonylmanganese(I), 98%

Click to view available options
Quantity:
1g
5g
14516-54-2
C5BrMnO5-
274.892
MFCD00049806
LOCZKKJCQVRJPQ-UHFFFAOYSA-M
bromopentacarbonylmanganese i,bromopentacarbonylmanganese i
10978692
carbon monoxide;manganese;bromide
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mn].[Br-]
14516-54-2
C5BrMnO5-
274.892
MFCD00049806
LOCZKKJCQVRJPQ-UHFFFAOYSA-M
bromopentacarbonylmanganese i,bromopentacarbonylmanganese i
10978692
carbon monoxide;manganese;bromide
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mn].[Br-]
Sensitivity Air & Moisture Sensitive
Color Orange
Quantity 1g
Linear Formula Mn(CO)5Br
UN Number UN3466
Formula Weight 274.89
Physical Form Crystalline
Percent Purity 98%
Chemical Name or Material Bromopentacarbonylmanganese(I)

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.