Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromophenol Blue, 0.1% (w/v) Aqueous Indicator Solution, For Acidity, APHA, Spectrum™ Chemical
Supplier: Spectrum Chemical Manufacturing Corporation B350125ML
Specifications
115-39-9, 7732-18-5, 34725-61-6 | |
Chemical Solution | |
Poly Bottle | |
125 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction