Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromophenol Blue, 0.1% w/v in denatured Ethanol, Reagents
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Bromophenol Blue, 0.1% w/v in denatured Ethanol |
CAS | 64-17-5, 67-63-0, 67-63-0 |
Boiling Point | 72°C |
Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
Percent Purity | 0.1% |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 0.8 |
Color | Orange/Red |
Quantity | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction