Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue, ACS Reagent (Sultone) not water soluble, Reagents

Catalog No. p-200021642
Click to view available options
Quantity:
1 g
5 g
10 g

Bromophenol Blue, ACS Reagent (Sultone) not water soluble, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Bromophenol Blue, ACS Reagent (Sultone) not water soluble
CAS 115-39-9
Molecular Formula C19H10Br4O5S
Synonym 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue
Molecular Weight (g/mol) 669.9607
Grade ACS
Packaging Glass Bottle
Specific Gravity 0.954
Color Orange
Quantity 10 g
Physical Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.