Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromophenol Blue sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals

Catalog Number AA38693AP
Click to view available options
Quantity:
100 mL
500 mL
34725-61-6
691.96
MFCD00013793
34725-61-6
691.96
MFCD00013793

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration 0.04% w/v aq. soln.
Merck Index 141444
Formula Weight 691.96
Color Red
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Bromophenol Blue sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit