Alfa Aesar™ Bromophenol Blue sodium salt, 0.04% w/v aq. soln.

Manufacturer: Alfa Aesar™ 38693M0

Quantity 6 x 100mL

 View more versions of this product

Catalog No. AA38693M0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34725-61-6
    Formula Weight 691.96
    MDL Number MFCD00013793
    Merck Index 14,1444
    Chemical Name or Material Bromophenol Blue sodium salt

    Specifications

    Formula Weight 691.96
    Quantity 6 x 100mL
    TSCA TSCA
    Concentration 0.04% w/v aq. soln.
    Physical Form Liquid
    MDL Number MFCD00013793
    CAS 34725-61-6
    Merck Index 14,1444
    Color Dark red
    Chemical Name or Material Bromophenol Blue sodium salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit