Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromophenol Blue TS 0.1% in dilute Alcohol, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Bromophenol Blue TS 0.1% in dilute Alcohol |
Boiling Point | 82°C |
CAS | 7732-18-5, 64-17-5, 64-17-5 |
Synonym | 3',3'',5',5''-Tetrabromosulfonphthalein, Tetrabromophenol blue, Bromphenol blue |
Grade | USP |
Packaging | Poly Bottle |
Specific Gravity | 0.9225 |
Color | Red |
Quantity | 120 mL |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction