Learn More
Selleck Chemical LLCSupplier Diversity Partner Bromopyruvic acid DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S542625MG
Bromopyruvic acid is a hexokinase II inhibitor with Ki of 2.4 mM for glycolysis/hexokinase inhibition. It is inhibitor of tumour cell energy metabolism and chemopotentiator of platinum drugs.
Catalog No. 50-194-8216