Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Bromostyrene, 95%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
98-81-7
C8H7Br
183.05
MFCD00012229
SRXJYTZCORKVNA-UHFFFAOYSA-N
1-bromovinyl benzene,alpha-bromostyrene,benzene, 1-bromoethenyl,1-bromostyrene,styrene, alpha-bromo,1-phenylvinyl bromide,alpha-bromostyrol russian,styrene, .alpha.-bromo,unii-lf0sj1821n,alpha-bromostyrol
66828
1-bromoethenylbenzene
C=C(C1=CC=CC=C1)Br
98-81-7
C8H7Br
183.05
MFCD00012229
SRXJYTZCORKVNA-UHFFFAOYSA-N
1-bromovinyl benzene,alpha-bromostyrene,benzene, 1-bromoethenyl,1-bromostyrene,styrene, alpha-bromo,1-phenylvinyl bromide,alpha-bromostyrol russian,styrene, .alpha.-bromo,unii-lf0sj1821n,alpha-bromostyrol
66828
1-bromoethenylbenzene
C=C(C1=CC=CC=C1)Br
Infrared Spectrum Authentic
Melting Point -44°C
Boiling Point 67°C to 70°C (4.0mmHg)
Flash Point 86°C
Specific Gravity 1.41
Refractive Index 1.5870 to 1.589
Density 1.4100g/mL
Stabilizer about 0.05% 3, 5-di-tert-butylcatechol
Quantity 100g
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula C6H5C(Br)=CH2
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Formula Weight 183.05
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material α-Bromostyrene, Stabilized
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-702-4