Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, 1% (w/v) Aqueous Indicator Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
34722-90-2,7732-18-5
C27H27Br2NaO5S
646.37
NMKFVGALBGZKGW-FKWCIMQXSA-M
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
34722-90-2,7732-18-5
C27H27Br2NaO5S
646.37
NMKFVGALBGZKGW-FKWCIMQXSA-M
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

B-177, 34722-90-2

Spectrum™ Bromothymol Blue, 1 Percent (w/v) Aqueous Indicator Solution is used as an acid-base indicator. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
CAS Max % 1%,99%
Packaging Poly Bottle
Quantity 1 L