Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromothymol Blue, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
5g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

76-59-5
C27H28Br2O5S
624.384
NUHCTOLBWMJMLX-UHFFFAOYSA-N
6450
CHEBI:86155

High quality chemicals for laboratory and specialized industrial use

CAS 76-59-5
Molecular Formula C27H28Br2O5S
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
Molecular Weight (g/mol) 624.384
PubChem CID 6450
ChEBI CHEBI:86155
Formula Weight 624.39
Packaging Wide Mouth Amber Glass Bottle
Quantity 5g
Chemical Name or Material Bromothymol Blue
Grade Baker Analyzed™ ACS Reagent
Show More Show Less