Bromothymol Blue sodium salt, 0.04% w/v aqueous solution, Alfa Aesar™

Manufacturer: Alfa Aesar 38701M0

Quantity 6 x 100mL

 View more versions of this product

Catalog No. AA38701M0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34722-90-2
    Physical Form Liquid
    Molecular Formula C27H27Br2NaO5S
    Formula Weight 646.38
    MDL Number MFCD00077263
    Chemical Name or Material Bromothymol Blue sodium salt

    Specifications

    Formula Weight 646.38
    Quantity 6 x 100mL
    Color Blue
    Physical Form Liquid
    Molecular Formula C27H27Br2NaO5S
    MDL Number MFCD00077263
    CAS 34722-90-2
    Odor Odorless
    Chemical Name or Material Bromothymol Blue sodium salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit