Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromotrimethylsilane, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
2857-97-8
C3H9BrSi
153.1
IYYIVELXUANFED-UHFFFAOYSA-N
trimethylsilyl bromide,trimethylbromosilane,silane, bromotrimethyl,trimethylsilicon bromide,tmsbr,trimethylsilylbromide,bromotrimethyl silane,tmbs,bromo trimethyl silane,tms bromide
76113
bromo(trimethyl)silane
C[Si](C)(C)Br
2857-97-8
C3H9BrSi
153.1
IYYIVELXUANFED-UHFFFAOYSA-N
trimethylsilyl bromide,trimethylbromosilane,silane, bromotrimethyl,trimethylsilicon bromide,tmsbr,trimethylsilylbromide,bromotrimethyl silane,tmbs,bromo trimethyl silane,tms bromide
76113
bromo(trimethyl)silane
C[Si](C)(C)Br
C-X bond formation reagent, Silylation reagent
Refractive Index 1.4200 to 1.4250
Color Colorless to Yellow
Specific Gravity 1.16
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 32°C
Quantity 100g
Density 1.1600g/mL
Melting Point -43.0°C
Boiling Point 79.0°C
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula (CH3)3SiBr
Fieser 09,73; 10,59; 12,79; 15,51; 16,50
Solubility Information Solubility in water: decomposes.
Formula Weight 153.1
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Bromotrimethylsilane
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 220-672-