Learn More
Bromotriphenylmethane, 98%, Thermo Scientific Chemicals
Description
It is considered as a more reactive reagent than Chlorotriphenylmethane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is considered as a more reactive reagent than Chlorotriphenylmethane, for the preparation of trityl esters. It is also used in one-pot conversion of amino acids to their N-trityl derivatives involving N,O-ditritylation and selective methanolysis of the trityl ester.
Solubility
Solubility in hot acetic acid is almost transparent.
Notes
Moisture sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 152°C to 156°C |
Density | 1.55 |
Boiling Point | 230°C (15 mmHg) |
Quantity | 50 g |
UN Number | UN1759 |
Beilstein | 1878494 |
Sensitivity | Moisture sensitive |
Formula Weight | 323.24 |
Percent Purity | 98% |
Chemical Name or Material | Bromotriphenylmethane |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 209-884-4 |
RTECSNumber | PA5550000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.