Alfa Aesar™ Bromotris(dimethylamino)phosphonium hexafluorophosphate, 98+%

Manufacturer: Alfa Aesar™ H2670803

Quantity 1g

 View more versions of this product

Catalog No. AAH2670803

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 50296-37-2
    Molecular Formula C6H18BrF6N3P2
    Formula Weight 388.07
    MDL Number MFCD00191864
    UN Number UN1759
    Synonym BroP
    Chemical Name or Material Bromotris(dimethylamino)phosphonium hexafluorophosphate

    Specifications

    Formula Weight 388.07
    Molecular Formula C6H18BrF6N3P2
    CAS 50296-37-2
    UN Number UN1759
    Melting Point >300°C
    Synonym BroP
    Quantity 1g
    Health Hazard 1 Danger
    Recommended Storage 2° to 8°C
    MDL Number MFCD00191864
    Health Hazard 2 GHS H Statement
    H350-H314-H318
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Chemical Name or Material Bromotris(dimethylamino)phosphonium hexafluorophosphate
    Assay ≥98%

    GHS H Statement
    H350-H314-H318
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    2° to 8°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit