Alfa Aesar™ Bromotris(triphenylphosphine)rhodium(I), 99.95% (metals basis), Rh 10.1% min

Hydrogenation

Manufacturer: Alfa Aesar™ 3982101

Quantity 0.25g

 View more versions of this product

Catalog No. AA3982101

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14973-89-8
    Formula Weight 969.69
    MDL Number MFCD00009828
    Chemical Name or Material Bromotris(triphenylphosphine)rhodium(I)

    Specifications

    Formula Weight 969.69
    Quantity 0.25g
    Physical Form Crystalline
    MDL Number MFCD00009828
    Concentration Rh 10.1% min.
    Solubility Information Soluble in chlorinated solvents
    CAS 14973-89-8
    Chemical Name or Material Bromotris(triphenylphosphine)rhodium(I)
    Assay 99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit