Bromthymol Blue-D pH Indicator Solution, pH 6.0 to 7.6, Lovibond™

Manufacturer: Tintometer Group 170S32OZ/PRC

Quantity 32 oz.

 View more versions of this product

Catalog No. 23-141762

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 34725-61-6
    Physical Form Liquid
    Synonym Bromothymol Blue
    Chemical Name or Material Bromthymol Blue-D pH Indicator Solution, pH 6.0 to 7.6

    Specifications

    Quantity 32 oz.
    Physical Form Liquid
    CAS 34725-61-6
    Synonym Bromothymol Blue
    Chemical Name or Material Bromthymol Blue-D pH Indicator Solution, pH 6.0 to 7.6

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit