Bronze Gauze, 20 mesh woven from 0.41mm (0.016 in.) in dia. wire, Alfa Aesar™

Manufacturer: Alfa Aesar 45144-GT

Quantity 150 x 150mm

 View more versions of this product

Catalog No. AA45144GT

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Gauze
    MDL Number MFCD00198187
    Chemical Name or Material Bronze

    Specifications

    Appearance Bronze colored gauze
    Odor Odorless
    Color Bronze
    Quantity 150 x 150mm
    Physical Form Gauze
    MDL Number MFCD00198187
    Chemical Name or Material Bronze

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit