Alfa Aesar™ Bronze gauze, alloy 220, 150 mesh woven from 0.066mm (0.0026in) in dia wire

Manufacturer: Alfa Aesar™ 45145GT

Quantity 150 x 150mm

 View more versions of this product

Catalog No. AA45145GT

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00198187
    Chemical Name or Material Bronze gauze, Alloy 220, 150 mesh woven from 0.066mm (0.0026 in.) dia. wire

    Specifications

    Quantity 150 x 150mm
    MDL Number MFCD00198187
    Mesh Size 150 Mesh
    Odor Odorless
    Chemical Name or Material Bronze gauze, Alloy 220, 150 mesh woven from 0.066mm (0.0026 in.) dia. wire

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit