Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bronze Gauze, Alloy 220, 40 mesh woven from 0.25mm (0.01 in.) in dia. wire, Alfa Aesar™

Typical composition of alloy: Cu 89.0-91.0%, Fe 0.05% max, Pb 0.5% max Zn remainder

Manufacturer:  Alfa Aesar 45166GT

Quantity 150 x 150mm

 View more versions of this product

Catalog No. AA45166GT


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Gauze, Alloy 220
MFCD00198187
Bronze

Specifications

150 x 150mm
Bronze colored gauze
Gauze, Alloy 220
Bronze
Bronze
Odorless
MFCD00198187
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit