Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Brorin/VWC2 Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2793855PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Brorin/VWC2.Specifications
7.81 pg/mL | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Brorin/VWC2. | |
Brorin/VWC2 | |
5 Plates | |
Sandwich ELISA | |
Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2 |
HRP | |
375567 | |
Antibody Pairs | |
7.81-500 pg/mL | |
Storage is content dependent. | |
Mouse | |
Human | |
VWC2 | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title