Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Brorin/VWC2 Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 7.81 pg/mL |
---|---|
Assay Sensitivity | 7.81-500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009809
|
Novus Biologicals™
NBP27938515PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Brorin/VWC2.Specifications
7.81 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2 | |
VWC2 | |
Storage is content dependent. |
7.81-500 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Brorin/VWC2. | |
Sandwich ELISA | |
375567 | |
Mouse | |
RUO | |
Brorin/VWC2 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title