Learn More
Sklar InstrumentsSupplier Diversity Partner Brown Needle Holder, Sklar, 6-3/4 in., Straight, Serrated, Stainless Steel. Sklar's Brown Needle Holder is a ratcheted, finger ring instrument used for holding needles. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 204867
Brown Needle Holder, Sklar, 6-3/4 in., Straight, Serrated, Stainless Steel. Sklar's Brown Needle Holder is a ratcheted, finger ring instrument used for holding needles. It is commonly used in plastic surgery and other procedures where suturing superficial tissue is necessary. This product is straight with rounded, serrated tips and a length of 6-3/4inches.
Catalog No. 50-143-7513