Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRP44L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584267
Description
Immunogen sequence: ATNEVAQLIQ GGRLIKHEMT KTA.
BRP44L (brain protein 44-like protein), also known as HSPC040 or CGI-129, is a 109 amino acid mitochondrial protein belonging to the UPF0041 family. The gene that encodes BRP44L maps to human chromosome 6. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
Specifications
BRP44L | |
Polyclonal | |
Unconjugated | |
MPC1 | |
0610006G08Rik; 3830411I18Rik; apoptosis-regulating basic protein; Arbp; Brain protein 44-like protein; Brp44l; CGI-129; dJ68L15.3; HSPC040; HSPC040 protein; mitochondrial pyruvate carrier 1; Mpc1; MPYCD; PNAS-115 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5U8 | |
MPC1 | |
Recombinant protein corresponding to Human MPC1. Recombinant protein control fragment (Product #RP-106663). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction