Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRP44L Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310637100UL
Description
BRP44L Polyclonal specifically detects BRP44L in Rat samples. It is validated for Western Blot.Specifications
BRP44L | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
brain protein 44-like, brain protein 44-like protein, CGI-129, dJ68L15.3, HSPC040 protein | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat BRP44L (NP_598245). Peptide sequence GALVRKAADYVRSKDFRDYLMSTHFWGPVANWGLPIAAINDMKKSPEIIS | |
100 μg | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
51660 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction