Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRP44L Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31063725UL

 View more versions of this product

Catalog No. NB126176


Add to cart

Description

Description

BRP44L Polyclonal antibody specifically detects BRP44L in Rat samples. It is validated for Western Blot
Specifications

Specifications

BRP44L
Unconjugated
PBS buffer, 2% sucrose
brain protein 44-like, brain protein 44-like protein, CGI-129, dJ68L15.3, HSPC040 protein
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat BRP44L (NP_598245). Peptide sequence GALVRKAADYVRSKDFRDYLMSTHFWGPVANWGLPIAAINDMKKSPEIIS
25 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Rat
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
51660
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit