Learn More
BRPF1 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00007862T01
Catalog No. 89-001-955
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_001003694.1
6.8
Transient overexpression cell Lysate was tested with Anti-BRPF1 antibody (H00007862-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
BRPF1 293T Cell Transient Overexpression Lysate(Denatured)
138.2
pCMV-BRPF1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.