Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRPF3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531848
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (94%), Rat (94%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Scaffold subunit of various histone acetyltransferase (HAT) complexes, such as the MOZ/MORF and HBO1 complexes, which have a histone H3 acetyltransferase activity (PubMed:16387653, PubMed:26620551, PubMed:26677226). Plays a role in DNA replication initiation by directing KAT7/HBO1 specificity towards histone H3 'Lys-14' acetylation (H3K14ac), thereby facilitating the activation of replication origins (PubMed:26620551). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity (PubMed:16387653). [UniProt].
Specifications
BRPF3 | |
Polyclonal | |
Unconjugated | |
BRPF3 | |
AI413466; AW493400; bromodomain and PHD finger containing 3; bromodomain and PHD finger containing, 3; bromodomain and PHD finger-containing protein 3; BRPF3; KIAA1286; mKIAA1286 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9ULD4 | |
BRPF3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 224 of Human BRPF3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction