Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRSK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112309
Description
BRSK1 Polyclonal Antibody for Western Blot, IP
BRSK1 (SAD1) is a serine/threonine kinase, and is a UV-induced DNA damage checkpoint kinase. BRSK1 is unbiquitously expressed, with highest levels of expression in the brain and testes. Similar to its yeast homolog, BRSK1 is thought to be involved in stress-induced cell cycle arrest. Overexpression of this protein leads to the G2/M arrest in HeLa S2 cells and UV-induced G2/M arrest could be partially abrogated by reduced expression of BRSK1 through the use of siRNA, indicating its role in DNA damage checkpoint function. More recently, it has been shown that both BRSK1 and the related protein BRSK2 are required for mammalian neuronal polarization. While BRSK1- and BRSK2-null mice were viable, double-mutant mice died within two hours of birth. Neurons from these mice showed uniformly-sized neurites as opposed to the normal long axon and multiple shorter dendrites. These neurites also displayed both axonal and dendritic markers. At least two isoforms of BRSK1 are known to exist.
Specifications
BRSK1 | |
Polyclonal | |
Unconjugated | |
Brsk1 | |
BR serine/threonine kinase 1; BR serine/threonine-protein kinase 1; Brain-selective kinase 1; brain-specific serine/threonine-protein kinase 1; Brsk1; Gm1100; hSAD1; KIAA1811; protein kinase SAD1A; RGD1563268; SAD1; SAD1 homolog; SAD1 kinase; SAD1A; SADB; SAD-B; SadB kinase short isoform; serine/threonine kinase SADB; serine/threonine kinase SAD-B; serine/threonine-protein kinase BRSK1; Serine/threonine-protein kinase SAD-B; synapses of Amphids Defective homolog 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
84446 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q8TDC3 | |
Brsk1 | |
E. coli-derived Human BRSK1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction