Learn More
Drg International IncSupplier Diversity Partner Brucella Igg Elisa DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA3455
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Brucella in serum and plasma. Brucella is a small Gram-negative bacterium (0.4-0.8 µm in diameter and 0.4-3.0 µm in length) which is non-flagellated, and non-spore-forming. Brucellosis is a nationally notifiable disease and reportable to the local health authority. The DRG Brucella IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Brucella abortus Antigen from strain S99-Weybridge. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Brucella-specific antibodies of positive specimens and controls are bound to the immobilized antigens.
Catalog No. 50-194-7560