Learn More
Drg International Inc DRG Brucella IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3456R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to Brucella in serum and plasma. This product is for Research Use Only. The DRG Brucella IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. Microtiter wells as a solid phase are coated with inactivated Brucella abortus Antigen from strain S99-Weybridge. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7561
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.