Learn More
BD Bruker MALDI Biotyper Sirius System
A purposefully engineered mass spectrometry instrument for the clinical microbiology laboratory connected with reliable antimicrobial susceptibility testing.
Supplier:  BD 1890133
Catalog No. B1890133
Request a Quote
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
Explore available promotions
This item is not returnable. View return policy
Description
Description
  • True benchtop solutions requiring less than 3 feet of counter space
  • FDA cleared library with 424 organisms in 333 groups covering anaerobes, Gram-positive bacteria, Gram negative bacteria, and yeast
  • Low-noise operating system
  • 110 V electrical requirements, with minimal heat output
  • Automated workflow enhancement and prep instruments available
  • Real time microbiology – no batching required with BD Bruker MALDI Biotyper workflow
  • Bruker Bacterial Test Standard (BTS) simplifies QC – easier and more convenient than other commercially available strains
  • Disposable target plates provide 96 spots and a unique barcode for full traceability, with no time consuming cleanup
  • The BD EpiCenter Data Management System's Plate Mapping module allows users to spot samples on MALDI target plates while simultaneously connecting them to appropriate BD Phoenix susceptibility panels
  • Straightforward result interpretation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mass Spectrometry Instrument
Bruker MALDI Biotyper Sirius System
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.