Learn More
Aviva Systems Biology Corp BRUNOL5 Antibody - middle region (ARP40970_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP40970_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRUNOL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BRUNOL5. Alternate names: BRUNOL-5, CELF5, CELF-5, BRUNOL5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62887