Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRUNOL6 (CELF6) Monoclonal Antibody (OTI1F8), OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811935


Description
BRUNOL6 (CELF6) Monoclonal antibody specifically detects BRUNOL6 (CELF6) in Human samples. It is validated for Western BlotSpecifications
BRUNOL6 (CELF6) | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96Q40 | |
Mouse | |
Protein A/G | |
RUO | |
65061 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI1F8 | |
Unconjugated | |
CDK15 | |
6330569O16Rik; Bruno -like 6, RNA binding protein; BRUNOL6; bruno-like 6, RNA binding protein; bruno-like protein 6; CELF6; CELF-6; CUG-BP and ETR-3 like factor 6; CUG-BP- and ETR-3-like factor 6; CUGBP Elav-like family member 6; CUGBP, Elav-like family member 6; RNA-binding protein BRUNOL-6 | |
Human recombinant protein fragment corresponding to amino acids 293-378 of human CDK15 (NP_631897) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction