Learn More
Sklar InstrumentsSupplier Diversity Partner Bruns Plaster Shears, Sklar, 9-1/4 in., Blunt/Blunt with Probe Tip, Curved, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 111295
Bruns Plaster Shears, Sklar, 9-1/4 in., Blunt/Blunt with Probe Tip, Curved, Serrated, Stainless Steel. Sklar Bruns Plaster Shears are non-ratcheted finger ring specialty scissors designed for use with plaster and fiberglass casting. These scissors should never be used on tissue. They are heavy shears with one larger ring for the thumb. The lower blade has heavy serrations that angle back towards the finger rings, and a blunted probe end so as not to cause injury to the patient while removing casting materials. It is also slightly longer than the standard blade. The standard blade tip is also blunt. This product has slightly curved blades with a 10cm stop peg in between the finger rings. It has overall length of 9-1/4 inches.
Catalog No. 50-143-7311