Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

BRWD1-IT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19159520UL

 View more versions of this product

Catalog No. NBP19159520



Description

Description

BRWD1-IT2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

BRWD1-IT2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
BRWD1 intronic transcript 2 (non-protein coding), C21orf87, NCRNA00257
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
Affinity Purified
BRWD1-IT2
Synthetic peptide directed towards the C terminal of human C21orf87. Peptide sequence LEARGLPCGARRTGPRRPVREMTLPSDPERATLPNPRLGAPAVPRRGPRS.
Immunogen affinity purified
RUO
Primary
257357
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit