Learn More
Akron Biotechnology LLCSupplier Diversity Partner BSA FRACTION V (PH 5.2) 500G DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK82450500
BSA, Fraction V (pH 5.2) U.S. Origin, is a protein that has typical applications in serum media, bacterial culture work and as a stabilizer for EIA and RIA enzymatic systems that require high purity. 500g
Catalog No. 50-753-3134