Learn More
Akron Biotechnology LLCSupplier Diversity Partner BSA FRACTION V (PH.7.0)500G DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK89050500
BSA, Fraction V (pH 7.0) U.S. Origin, is a protein that has typical applications in serum media, bacterial culture work and as a stabilizer for EIA and RIA enzymatic systems that require high purity. 500g
Catalog No. 50-753-3132