Learn More
Gemini Bio ProductsSupplier Diversity Partner BSA V FATTY ACID FREE 20G DFS Item
Manufacturer:  Gemini Bio ProductsSupplier Diversity Partner 700107P/20
A highly purified, lyophilized powder, this is processed to provide a minimum of 98% albumin and to be essentially free of non-esterified fatty acids. The pH is adjusted to 7.0. It's used as a blocking reagent in EIA, RIA and ELISA systems.
Catalog No. 50-753-3072