Learn More
Gemini Bio ProductsSupplier Diversity Partner BSA V - PROTEASE FREE 20G DFS Item
Manufacturer:  Gemini Bio ProductsSupplier Diversity Partner 700101P/20
Gemini's protease-free BSA is a highly purified, lyophilized powder. This product is processed to provide a minimum of 98% albumin and to be essentially free of detectable protease. The pH is adjusted to 7.0.
Catalog No. 50-753-3054