Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BSG Mouse anti-Human, Clone: 56.3_1D5, Abnova™

Mouse monoclonal antibody raised against human BSG.

Manufacturer:  Abnova Corporation MAB12754

Catalog No. 89-933-671



Description

Description

The protein encoded by this gene is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. The encoded protein is also a member of the immunoglobulin superfamily. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

BSG
56.3_1D5
Mouse monoclonal antibody raised against human BSG.
In PBS (0.02% sodium azide).
5F7/CD147/EMMPRIN/M6/OK/TCSF
Mouse
IgG2b
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunocytochemistry (10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
BSG
Human Myeloid Leukemia cells (HL60)
Protein A/G purification
RUO
Primary
682
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit