Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTBD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139775
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PSPSSLGPLL PLQREPLYNW QATKASLKER FAFLFNSELL SDVRFVLGKG.
The C-terminus of BTBD1 binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
BTBD1 | |
Polyclonal | |
Unconjugated | |
BTBD1 | |
1190005H08Rik; BTB (POZ) domain containing 1; BTB domain containing 1; BTB/POZ domain-containing protein 1; Btbd1; C15orf1; glucose signal-repressing protein; Gsrp; HCV NS5A-transactivated protein 8; hepatitis C virus NS5A-transactivated protein 8; NS5ATP8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0C5 | |
BTBD1 | |
Recombinant protein corresponding to Human BTBD1. Recombinant protein control fragment (Product #RP-108750). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction