Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTBD17 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548250

Catalog No. PA548250

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BTBD17
Polyclonal
PBS with 0.09% sodium azide
A6NE02, Q9DB72
BTB/POZ domain-containing protein 17, Galectin-3-binding protein-like, BTBD17
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
388419, 72014
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BTBD17
Liquid
BTBD17
KLH conjugated synthetic peptide between 411-440 amino acids from the C-terminal region of human BTBD17.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit