Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BTBD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559793
Description
Immunogen sequence: ILDAIKVRSE SRDMDLNYRG MLIPEENIAT MKYGAQVVKG ELKSALLDGD TQNYDLDHGF SRHPIDDDCR SGIEIKLGQP SIINHIRILL WDR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This locus encodes a BTB/POZ domain-containing protein. This domain is known to be involved in protein-protein interactions. Polymorphisms at this locus have been reported to be associated with susceptibility to Restless Legs Syndrome and may also be associated with Tourette Syndrome.
Specifications
BTBD9 | |
Polyclonal | |
Unconjugated | |
BTBD9 | |
1700023F20Rik; 4930402L05; AI464421; BTB (POZ) domain containing 9; BTB domain containing 9; BTB/POZ domain-containing protein 9; Btbd9; dJ322I12.1; KIAA1880 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96Q07 | |
BTBD9 | |
Recombinant protein corresponding to Human BTBD9. Recombinant protein control fragment (Product #RP-95020). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction