Tocris™ BTC

Selective estrogen receptor modulator (SERM)

Manufacturer: Tocris™ 5421/50

 View more versions of this product

Catalog No. 54-215-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors BTC
    Purity >99
    Molecular Weight 242.29
    Quantity 50mg
    Product Type BTC

    Selective estrogen receptor modulator (SERM); induces regression of tamoxifen-resistant, hormone independent xenograft tumors. Displays no significant gain in uterine weight in treated mice.

    • Chemical Name: 2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol
    • Formula: C14H10O2S
    • CAS Number: 63676-22-2
    • Solubility: Soluble to 50mM in DMSO and to 50mM in ethanol
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit