BTCP maleate, Tocris

Potent dopamine uptake inhibitor

Manufacturer: R&D Systems 0702/50

Quantity 50mg

 View more versions of this product

Catalog No. 07-025-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Solid

    Specifications

    Quantity 50mg
    Physical Form Solid
    Formula Weight 415.55
    Assay Percent Range >98%
    Molecular Formula C19H25NS·C4H4O4
    CAS 207455-25-2
    Synonym 1-[1-(2-Benzo[b]thienyl)cyclohexyl)]piperidine maleate
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material BTCP maleate

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit