Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTEX Solution, MilliporeSigma™ Supelco™
BTEX are important chemicals, which are commonly used as chemical intermediates and solvents in the production of plastics.{11)
$91.00 - $241.00
Specifications
CAS | 108-88-3 |
---|---|
Type | Analytical Standard |
Grade | Analytical Standard |
Density | 1.099 g/mL (at 20°C) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-100-2693
|
MilliporeSigma Supelco
437281ML |
1 mL |
Each for $91.00
|
|
|||||
11-100-2692
|
MilliporeSigma Supelco
4372810X1ML |
10 x 1 mL |
Each for $241.00
|
|
|||||
Description
EnvironmentalSpecifications
108-88-3 | |
Analytical Standard | |
Analytical Standard | |
1.099 g/mL (at 20°C) |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title