Learn More
Aviva Systems Biology Corp BTF3L3 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP58359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTF3L3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BTF3L3. Alternate names: BTF3L3, HUMBTFDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62292
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.