Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTG1 Antibody (OTI7H6) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270286

Catalog No. NBP270286


Add to Cart

Description

Description

BTG1 Monoclonal specifically detects BTG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

BTG1
Monoclonal
LYOPH
Immunohistochemistry 1:150
B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1
Mouse
19 kDa
100 μg
694
Human, Mouse, Rat
Lyophilized
Immunohistochemistry
OTI7H6
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
BTG1
Full length human recombinant protein of human BTG1 (NP_001722) produced in E.coli.
Affinity Purified
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit